Human Recombinant Ephrin-A1
Supplier: Bon Opus Biosciences
CA70-10UG CA70-1MG CA70-500UG CA70-50UG
102863-734EA 118.46 USD
102863-734 102863-736 102863-738 102863-740
Human Recombinant Ephrin-A1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Ephrin-A1 is produced by our Mammalian expression system and the target gene encoding Asp19-Ser182 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.