Human Recombinant Cadherin 11 Type 2 OB-cadherin (Osteoblast)
Supplier: Bon Opus Biosciences
CA45-10UG CA45-1MG CA45-500UG CA45-50UG
102863-662EA 118.46 USD
102863-662 102863-664 102863-666 102863-668
Human Recombinant Cadherin 11 Type 2 OB-cadherin (Osteoblast)
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Cadherin-11 is produced by our Mammalian expression system and the target gene encoding Phe23-Thr617 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.