Human Recombinant Leucine-Rich Repeat Transmembrane Protein FLRT3
Supplier: Bon Opus Biosciences
C970-10UG C970-1MG C970-500UG C970-50UG
102863-502EA 118.46 USD
102863-502 102863-504 102863-506 102863-508
Human Recombinant Leucine-Rich Repeat Transmembrane Protein FLRT3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human FLRT3 is produced by our Mammalian expression system and the target gene encoding Lys29-Pro528 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.