Human Recombinant Sorbitol Dehydrogenase
Supplier: Bon Opus Biosciences
C885-10UG C885-1MG C885-500UG C885-50UG
102863-470EA 170.15 USD
102863-470 102863-472 102863-474 102863-476
Human Recombinant Sorbitol Dehydrogenase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Sorbitol Dehydrogenase is produced by our Mammalian expression system and the target gene encoding Ala2-Pro357 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.