Human Recombinant Protein S100-A9
Supplier: Bon Opus Biosciences
C795-10UG C795-50UG C795-500UG C795-1MG
102943-002EA 57.08 USD
102943-002 102943-004 102943-006 102943-008
Human Recombinant Protein S100-A9
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human S100 Calcium Binding Protein A9 is produced by our E.coli expression system and the target gene encoding Thr2-Pro114 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.