Mouse Recombinant Cxcl12
Supplier: Bon Opus Biosciences
C698-10UG C698-50UG C698-500UG C698-1MG
102942-666EA 118.46 USD
102942-666 102942-668 102942-670 102942-672
Mouse Recombinant Cxcl12
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse C-X-C motif chemokine 12 is produced by our E.coli expression system and the target gene encoding Lys22-Lys89 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.