Recombinant Tumor Necrosis Factor Receptor Superfamily Member 1b
Supplier: Bon Opus Biosciences
C694-10UG C694-50UG C694-500UG C694-1MG
102942-634EA 107.69 USD
102942-634 102942-636 102942-638 102942-640
Recombinant Tumor Necrosis Factor Receptor Superfamily Member 1b
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Tumor Necrosis Factor Receptor II is produced by our Mammalian expression system and the target gene encoding Val23-Gly258 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.