Human Recombinant Kunitz-Type Protease Inhibitor 2
Supplier: Bon Opus Biosciences
C644-10UG C644-1MG C644-500UG C644-50UG
102863-342EA 118.46 USD
102863-342 102863-344 102863-346 102863-348
Human Recombinant Kunitz-Type Protease Inhibitor 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Hepatocyte Growth Factor Activator Inhibitor Type 2 is produced by our Mammalian expression system and the target gene encoding Ala28-Lys197 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.