Human Recombinant B-Lymphocyte Antigen CD19
Supplier: Bon Opus Biosciences
C572-10UG C572-50UG C572-500UG C572-1MG
102942-498EA 118.46 USD
102942-498 102942-500 102942-502 102942-504
Human Recombinant B-Lymphocyte Antigen CD19
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CD19 is produced by our Mammalian expression system and the target gene encoding Pro20-Lys291 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.