Human Recombinant Plexin Domain-Containing Protein 1
Supplier: Bon Opus Biosciences
C529-10UG C529-1MG C529-500UG C529-50UG
102862-790EA 118.46 USD
102862-790 102862-792 102862-794 102862-796
Human Recombinant Plexin Domain-Containing Protein 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Plexin Domain-Containing Protein 1 is produced by our Mammalian expression system and the target gene encoding Leu19-Thr426 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.