Human Recombinant Oxidized Low-Density Lipoprotein Receptor 1
Supplier: Bon Opus Biosciences
C524-10UG C524-1MG C524-500UG C524-50UG
102862-750EA 90.46 USD
102862-750 102862-752 102862-754 102862-756
Human Recombinant Oxidized Low-Density Lipoprotein Receptor 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human LOX-1 is produced by our Mammalian expression system and the target gene encoding Ser61-Gln273 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.