Human Recombinant Chymotrypsin-C
Supplier: Bon Opus Biosciences
C518-10UG C518-1MG C518-500UG C518-50UG
102862-710EA 170.15 USD
102862-710 102862-712 102862-714 102862-716
Human Recombinant Chymotrypsin-C
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Chymotrypsin-C is produced by our Mammalian expression system and the target gene encoding Cys17-Leu268 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.