Human Recombinant MHC Class I Polypeptide-Related Sequence A
Supplier: Bon Opus Biosciences
C489-10UG C489-1MG C489-500UG C489-50UG
102862-518EA 79.69 USD
102862-518 102862-520 102862-522 102862-524
Human Recombinant MHC Class I Polypeptide-Related Sequence A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human MHC Class I-related Protein A is produced by our Mammalian expression system and the target gene encoding Glu24-Gln308 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.