Human Recombinant Mannose-Binding Protein C
Supplier: Bon Opus Biosciences
C488-10UG C488-1MG C488-500UG C488-50UG
102862-510EA 90.46 USD
102862-510 102862-512 102862-514 102862-516
Human Recombinant Mannose-Binding Protein C
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Mannose Binding Lectin 2 is produced by our Mammalian expression system and the target gene encoding Glu21-Ile248 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.