Human Recombinant WARS also known as Tryptophanyl-tRNA synthetase
Supplier: Bon Opus Biosciences
C300-10UG C300-1MG C300-500UG C300-50UG
102509-212EA 107.69 USD
102509-212 102509-214 102509-216 102509-218
Human Recombinant WARS also known as Tryptophanyl-tRNA synthetase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human TrpRS is produced by our E.coli expression system and the target gene encoding Met1-Gln471 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.