Human Recombinant MOB Kinase Activator 1A
Supplier: Bon Opus Biosciences
C297-10UG C297-1MG C297-500UG C297-50UG
102861-182EA 170.15 USD
102861-182 102861-184 102861-186 102861-188
Human Recombinant MOB Kinase Activator 1A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human MOB Kinase Activator 1A is produced by our E.coli expression system and the target gene encoding Ser2-Arg216 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.