Human Recombinant Protein SCO1 Homolog Mitochondrial
Supplier: Bon Opus Biosciences
C259-10UG C259-1MG C259-500UG C259-50UG
102860-878EA 170.15 USD
102860-878 102860-880 102860-882 102860-884
Human Recombinant Protein SCO1 Homolog Mitochondrial
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Protein SCO1 Homolog Mitochondrial is produced by our E.coli expression system and the target gene encoding Gly132-Ser300 is expressed with a GST tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.