Human Recombinant Fibroblast Growth Factor 21
Supplier: Bon Opus Biosciences
C223-10UG C223-1MG C223-500UG C223-50UG
102860-590EA 90.46 USD
102860-590 102860-592 102860-594 102860-596
Human Recombinant Fibroblast Growth Factor 21
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Fibroblast Growth Factor 21 is produced by our E.coli expression system and the target gene encoding His29-Ser209 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.