Human Recombinant Annexin A6
Supplier: Bon Opus Biosciences
C207-10UG C207-1MG C207-500UG C207-50UG
102860-462EA 90.46 USD
102860-462 102860-464 102860-466 102860-468
Human Recombinant Annexin A6
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Annexin A6 is produced by our E.coli expression system and the target gene encoding Ala2-Asp673 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.