Human Recombinant Small Ubiquitin-Related Modifier 2
Supplier: Bon Opus Biosciences
C176-10UG C176-1MG C176-500UG C176-50UG
102860-214EA 11.85 USD
102860-214 102860-216 102860-218 102860-220
Human Recombinant Small Ubiquitin-Related Modifier 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SUMO2 is produced by our E.coli expression system and the target gene encoding Met1-Gly93 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.