Human Recombinant 26S Proteasome Non-ATPase Regulatory Subunit 10
Supplier: Bon Opus Biosciences
C166-10UG C166-1MG C166-500UG C166-50UG
102860-134EA 118.46 USD
102860-134 102860-136 102860-138 102860-140
Human Recombinant 26S Proteasome Non-ATPase Regulatory Subunit 10
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Gankyrin is produced by our E.coli expression system and the target gene encoding Met1-Gly226 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.