Human Recombinant Protein Phosphatase 1 Regulatory Subunit 1A

Supplier: Bon Opus Biosciences

C163-10UG C163-1MG C163-500UG C163-50UG
102860-110EA 118.46 USD
102860-110 102860-112 102860-114 102860-116
Human Recombinant Protein Phosphatase 1 Regulatory Subunit 1A
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human PPP1R1A is produced by our E.coli expression system and the target gene encoding Met1-Val171 is expressed with a 6His tag at the C-terminus.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR