Human Recombinant Fatty Acid-Binding Protein Adipocyte
Supplier: Bon Opus Biosciences
C136-10UG C136-1MG C136-500UG C136-50UG
102859-910EA 118.46 USD
102859-910 102859-912 102859-914 102859-916
Human Recombinant Fatty Acid-Binding Protein Adipocyte
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human FABP4 is produced by our E.coli expression system and the target gene encoding Cys2-Ala132 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.