Human Recombinant Stromal Cell-Derived Factor 1
Supplier: Bon Opus Biosciences
C121-10UG C121-1MG C121-500UG C121-50UG
102859-790EA 170.15 USD
102859-790 102859-792 102859-794 102859-796
Human Recombinant Stromal Cell-Derived Factor 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-X-C Motif Chemokine 12 is produced by our E.coli expression system and the target gene encoding Lys22-Lys89 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.