Human Recombinant DNA-(Apurinic or Apyrimidinic Site) Lyase
Supplier: Bon Opus Biosciences
C101-10UG C101-1MG C101-500UG C101-50UG
102859-630EA 118.46 USD
102859-630 102859-632 102859-634 102859-636
Human Recombinant DNA-(Apurinic or Apyrimidinic Site) Lyase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Apurinic-Apyrimidinic Endonuclease 1 is produced by our E.coli expression system and the target gene encoding Pro2-Leu318 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.