Human Recombinant Ciliary Neurotrophic Factor
Supplier: Bon Opus Biosciences
C098-10UG C098-1MG C098-500UG C098-50UG
102859-606EA 132.46 USD
102859-606 102859-608 102859-610 102859-612
Human Recombinant Ciliary Neurotrophic Factor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CNTF is produced by our E.coli expression system and the target gene encoding Ala2-Met200 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.