Human Recombinant Leptin
Supplier: Bon Opus Biosciences
C067-10UG C067-1MG C067-500UG C067-50UG
102859-398EA 28 USD
102859-398 102859-400 102859-402 102859-404
Human Recombinant Leptin
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Leptin is produced by our E.coli expression system and the target gene encoding Val22-Cys167 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.