Human Recombinant C-C Motif Chemokine 5
Supplier: Bon Opus Biosciences
C062-10UG C062-1MG C062-500UG C062-50UG
102859-366EA 132.46 USD
102859-366 102859-368 102859-370 102859-372
Human Recombinant C-C Motif Chemokine 5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-C Motif Chemokine 5 is produced by our E.coli expression system and the target gene encoding Ser24-Ser91 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.