Human Recombinant Myelin-Oligodendrocyte Glycoprotein
Supplier: Bon Opus Biosciences
C051-10UG C051-1MG C051-500UG C051-50UG
102859-302EA 79.69 USD
102859-302 102859-304 102859-306 102859-308
Human Recombinant Myelin-Oligodendrocyte Glycoprotein
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Myelin Oligodendrocyte Glycoprotein is produced by our E.coli expression system and the target gene encoding Gly30-Gly154 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.