Mouse Recombinant Fibroblast Growth Factor 1
Supplier: Bon Opus Biosciences
C043-10UG C043-1MG C043-500UG C043-50UG
102859-238EA 57.08 USD
102859-238 102859-240 102859-242 102859-244
Mouse Recombinant Fibroblast Growth Factor 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Fibroblast growth factor 1 is produced by our E.coli expression system and the target gene encoding Phe16-Asp155 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.