Human Recombinant CD40 Ligand
Supplier: Bon Opus Biosciences
C011-10UG C011-1MG C011-500UG C011-50UG
102859-054EA 57.08 USD
102859-054 102859-056 102859-058 102859-060
Human Recombinant CD40 Ligand
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human TNFSF5 is produced by our E.coli expression system and the target gene encoding Met113-Leu261 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.