Palbociclib isethionate ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 2-hydroxyethanesulphonate, 6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 2-hydroxyethanesulfonate

CHM234-0100 CHM234-0200 CHM234-0500 CHM234-1000 CHM234-2000
76203-334EA 170.04 USD
76203-334 76203-336 76203-338 76203-340 76203-342 76203-344
Palbociclib isethionate ≥99% (by HPLC)
Palbociclib isethionate

Palbociclib, also known as PD0332991, is an orally available pyridopyrimidine-derived cyclin-dependent kinase (CDK) inhibitor with potential antineoplastic activity.


  • CDK inhibitor


Yellow solid.


Soluble in DMSO.

Formula: C₂₄H₂₉N₇O₂·C₂H₆O₄S
Storage Temperature: Freezer
MDL Number: MFCD22666589
CAS Number: 827022-33-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eeed8a14457e9352792acb2e7966f6510dac9cf9&vid=b3451661a9df93cc015d23743e71adab1b93986f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR