Palbociclib free base ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one

CHM232-0050 CHM232-0100 CHM232-0200 CHM232-0500 CHM232-1000 CHM232-2000
76203-318EA 160.23 USD
76203-318 76203-320 76203-322 76203-324 76203-326 76203-328
Palbociclib free base ≥99% (by HPLC)
Palbociclib

Palbociclib, also known as PD0332991, is an orally available pyridopyrimidine-derived cyclin-dependent kinase (CDK) inhibitor with potential antineoplastic activity.


  • CDK inhibitor


Yellow solid.


Soluble in DMSO.

Formula: C₂₄H₂₉N₇O₂
Storage Temperature: Freezer
MDL Number: MFCD11840850
CAS Number: 571190-30-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e1e4ebc3de695f4de6be66b9faf221ed1ef9b31&vid=973a11a725576564107140a0fc7fcf902e5ea53f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR