IL-1β (rat), ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-131A
76199-136EA 834 USD
76199-136
IL-1β (rat), ELISA kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of IL-1β in culture supernatants and serum of rat origin.
  • Sensitive - use less precious sample per test
  • Higher throughput - test up to 40 samples in duplicate in less than 4 hours
  • Quantitative - obtain statistically significant results and compare data between multiple experiments
  • Ready-to-use - pre-coated plates save time and reduce errors
The IL-1β (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 4 hours.
Ordering information: Contains Microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, Biotinylated antibody, Standard, Standard diluent, Wash buffer concentrate, TMB Substrate, Stop solution.