Bradford Protein Assay Kit (B-Pak), OZ Biosciences
Supplier: OZ Biosciences
BA00050
BA00070
BA00100
103258-314EA
147.69
USD
103258-314
103258-316
103258-318
Bradford Protein Assay Kit (B-Pak), OZ Biosciences
Assays
Protein Assays
Bradford Protein Assay Kit (B-Pak) is a straightforward and rapid kit for determining the concentration of proteins in solution.This ready-to-use kit is provided with 1X reagent & 2 sets of BSA standards which means that no dilution, filtration or calculation are required
- Accurate determination of protein concentration
- Ready to use prediluted standard protein
- Cheaper than competitors
- Convenient packaging
- Fast and easy-to-use Bradford protein assay
Simply process your assay in a few minutes. B-Pak is provided with adapted protocols for all experimental conditions: micro and macroassays.
Delivery information: Shipped at room temperature
Packaging: Store at +4°C