Pep-1: Chariot Peptide

Supplier: AnaSpec
AS-61253
103006-480EA 314.23 USD
103006-480
Pep-1: Chariot Peptide
Proteins and Peptides
Pep-1 is one of the synthetic cell-penetrating peptides (CPPs), which has been successfully used to deliver a variety of proteins and other biopharmaceutical macromolecules into cells in a non-disruptive way. It is a CPP with primary amphipathicity (i.e amphipathicity resulting from the amino acid sequence itself, not from the folding structure) that comprises a tryptophan-rich so-called ‘hydrophobic’ domain, a hydrophilic domain derived from an NLS (nuclear localization signal) of SV40 (simian virus 40) large T-antigen, and a spacer between them.
Sequence:KETWWETWWTEWSQPKKKRKV
MW:2848.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR