Insulin Receptor (1142-1153)

Supplier: AnaSpec
AS-20274
102998-160EA 442.85 USD
102998-160
Insulin Receptor (1142-1153)
Proteins and Peptides
Peptides are used as insulin receptor tyrosine kinase substrates.
Sequence:TRDIYETD-pY-YRK
MW:1702.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR