[Lys(Me2)36]-Histone H3 (31-41)

Supplier: AnaSpec
AS-64380-1
103008-038EA 314.23 USD
103008-038
[Lys(Me2)36]-Histone H3 (31-41)
Proteins and Peptides
This peptide is Histone H3 amino acid residues 31 to 41 di-methylated at Lys-36.
Sequence:STGGV-K(Me2)-KPHRY
MW:1257.4 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR