[Lys(Me2)27, Lys(Me1)36]-Histone H3 (21-44)-GK,Biotin

Supplier: AnaSpec
AS-65449-1
103009-918EA 342 USD
103009-918
[Lys(Me2)27, Lys(Me1)36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This peptide is Histone 3, with amino acid residues 21 to 44. It is dimethylated at Lys27 and monomethylated Lys36, with a C-terminal G linker followed by a biotinylated lysine. Histone methylation plays an important role in the regulation of chromatin structure and function. 
Sequence:ATKAAR-K(Me2)-SAPATGGV-K(Me1)-KPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR