[Lys(Me1)27]-Histone H3 (21-44)-GK,Biotin

Supplier: AnaSpec
AS-64365-1 AS-64365-025
103008-002EA 599.23 USD
103008-002 103008-000
[Lys(Me1)27]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 21 to 44 mono-methylated at Lys-27 with an additional C-terminal glycine followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAAR-K(Me1)-SAPATGGVKKPHRYRPG-GK(Biotin)
MW:2931.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR