[Lys(Me1)23]-Histone H3 (21-44)-GK,Biotin

Supplier: AnaSpec
AS-64617-1
103008-302EA 599.23 USD
103008-302
[Lys(Me1)23]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 21 to 43. It is monomethylated at lysine 23 with a C-terminal G linker followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:AT-K(Me1)-AARKSAPSTGGVKKPHRYRPG-GK(Biotin)-NH2
MW:2946.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR