Influenza Virus Control Peptide Pool

Supplier: AnaSpec
AS-62340
103007-298EA 764.38 USD
103007-298
Influenza Virus Control Peptide Pool
Proteins and Peptides
These 12 Influenza virus peptides, at 0.25 mg each (total of 3 mg/vial), constitute part of the CEF Control Peptide Pool (cat# 61036-025). Now available separately as Influenza Control Peptide Pool, EBV Control Peptide Pool (cat# 62341) and CMV Control Peptide Pool (cat# 62339), these peptides have been used in the stimulation of IFNγ release from CD8+ T cells in individuals with defined HLA types, these epitopes are useful in applications such as ELISPOT, intracellular cytokine and CTL assays. All peptides are provided as net weight based on peptide content.
Sequence:
MW:
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR