HSV-gB2 (498-505)

Supplier: AnaSpec
AS-61708
103006-920EA 138.85 USD
103006-920
HSV-gB2 (498-505)
Proteins and Peptides
This is the immunodominant epitope gB-8p from herpes simplex virus (HSV) glycoprotein B (gB), amino acids 498 to 505.
Sequence:SSIEFARL
MW:922.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR