Magainin 2

Supplier: AnaSpec
AS-20639
102996-070EA 185.62 USD
102996-070
Magainin 2
Proteins and Peptides
Magainin 2 assumes an amphiphilic helix when bound to acidic phospholipids, forming a pore composed of a dynamic, peptide-lipid supramolecular complex.
Sequence:GIGKFLHSAKKFGKAFVGEIMNS
MW:2466.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR