Tau peptide,Biotin

Supplier: AnaSpec
AS-65409-1
103009-512EA 691.31 USD
103009-512
Tau peptide,Biotin
Proteins and Peptides
The microtubule-associated protein Tau, whose hyperphosphorylated form is associated to the Alzheimer's disease, is also known to be post-translationally modified by the addition of N-acetyl-D-glucosamine to some Ser or Thr. The Ser-400 tau O-GlcNAc modification was detected in rat brain.
Sequence:KWKHGAEIVYKSPVV-S(OGlcNAc)-GDTSPRHLSNVK-K(biotin)-NH2
MW:3677.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR