Cyclo (-RADfK-), RGD Negative Control

Supplier: AnaSpec
AS-62351
103007-308EA 206.08 USD
103007-308
Cyclo (-RADfK-), RGD Negative Control
Proteins and Peptides
This peptide is a negative control for the cyclo (-RGDfK), the RGD peptide. RGD peptides are modulators of cell adhesion and are recognized by several members of the integrin family. This peptide has low affinity binding to integrin peptides.
Sequence:Cyclo(-RADfK-)
MW:617.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR