Hyaluronan-Binding Peptide

Supplier: AnaSpec
AS-65199
103009-026EA 255.77 USD
103009-026
Hyaluronan-Binding Peptide
Proteins and Peptides
This peptide is a hyaluronan-binding peptide biotinylated through a C-terminal GGGSK linker. Hyaluronan (HA) is a nonsulfated glycosaminoglycan expressed in the extracellular matrix and on cell surfaces. HA plays a role in fertilization, embryonic development, wound healing, angiogenesis, leukocyte trafficking to inflamed tissues, and cancer metastasis. This peptide has been shown to block HA binding to CD44 receptors and inhibit T cell proliferation.
Sequence:GAHWQFNALTVRGGGS-K(Biotin)
MW:2012.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR