Human Leptin (93-105)

Supplier: AnaSpec
AS-62853
103007-536EA 160.77 USD
103007-536
Human Leptin (93-105)
Proteins and Peptides
This is amino acids 93 to 105 fragment of human leptin (anti-obesity protein). Leptin is a 167-amino acid plasma protein that is synthesized in adipose tissue and acts as a blood-borne hormone responsible for weight maintenance.
Sequence: NVIQISNDLENLR
MW: 1527.7 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR