Human Cholecystokinin (26-33)

Supplier: AnaSpec
AS-20741
102996-106EA 353.69 USD
102996-106
Human Cholecystokinin (26-33)
Proteins and Peptides
C-terminal sulfated and amidated octapeptide Cholecystokinin (sulfated CCK-8) has the full biological action of the full-length 33-amino acid long Cholecystokinin (CCK). CCK acts both as a hormone and a neurotransmitter and is found in the GI system and the central nervous system. It is a satiety peptide that inhibits food intake.
Sequence: D-Y(SO3H)-MGWMDF-NH2
MW: 1143.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR