Rat Renin FRET Substrate

Supplier: AnaSpec
AS-62334-01
103007-290EA 394.62 USD
103007-290
Rat Renin FRET Substrate
Proteins and Peptides
This renin FRET peptide is a specific substrate for rat renin. In the FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage into two separate fragments by rat renin, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission = 490/520 nm. The SensoLyte® 520 Renin Assay Kit (cat # 72040) contains the human renin FRET substrate.
MW: 2000 - 2200 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR